Would you like to log in again?
Mixer, Sulzer 24 Element Quadro, ELLSWORTH ADHESIVES CUST SERV TE
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
300-026
76240-826CS 2413.67 USD InStock
76240-826
Mixer, Sulzer 24 Element Quadro, ELLSWORTH ADHESIVES CUST SERV TE
Tool Sets
Static mixer is used to mix and dispense adhesive from 50 ml AA series cartridges - both 1:1 and 2:1 ratios.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty