Would you like to log in again?
2xYT-Broth (Capsules), MP Biomedicals
Supplier: MP Biomedicals
113012031
76020-266EA 329.05 USD InStock
76020-266
2xYT-Broth (Capsules), MP Biomedicals
Cell Culture Media
Sterile YT media formulations are optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages.
  • 31 g (31 capsules or 6 large capsules) per liter
  • Contents/l: 16 g, tryptone, 10 g yeast extract, 5 g NaCl bacterial growth medium
  • Medium optimized for the growth and maintenance of M13 or other filamentous phage
Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in 2X YT Broth formulations. Chosen reagents are combined in the accurate proportions according to the formula listed above and thoroughly blended for a uniform distribution. A sample of the blended formulation was used to prepare liquid. The combinations were mixed to dissolve components and then autoclaved at 121 °C for 15 minutes.
2X YT-Broth is a nutritional rich medium providing nitrogen and growth factors optimized for the growth of recombinant E. coli strains infected with the M13 bacteriophage or other filamentous single stranded DNA bacteriophages thus allowing the bacteriophage to reproduce in large quantities without exhausting the host cells.
YT media formulations are optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages.
Add 31 capsules per liter of purified water and autoclave at 121 °C for 15 minutes.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis.
Certifications: Molecular Biology Certified Bacterial Growth Medium, Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty