Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2xYT Broth (Large Capsules), MP Biomedicals

Supplier: MP Biomedicals
113012036
76202-284EA 522.92 USD InStock
76202-284
2xYT Broth (Large Capsules), MP Biomedicals
Cell Culture Media
2XYT-Broth is a nutritional rich medium providing nitrogen and growth factors optimized for the growth of recombinant E. coli strains infected with the M13 bacteriophage or other filamentous single stranded DNA bacteriophages thus allowing the bacteriophage to reproduce in large quantities without exhausting the host cells.

  • 31 g (31 capsules or 6 large capsules) per liter
  • Contents/l: 16 g, tryptone, 10 g yeast extract, 5 g NaCl bacterial growth medium
  • Medium optimized for the growth and maintenance of M13 or other filamentous phage
  • Autoclave at 121 °C for 15 minutes

YT media formulations are optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages.

Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis.

Certifications: Molecular Biology Certified bacterial growth medium powder.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty