Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terrific Broth, MP Biomedicals

Supplier: MP Biomedicals
113045032
IC113045032EA 460.04 USD InStock
IC113045032
Terrific Broth, MP Biomedicals
Cell Culture Media
Rich Medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.

  • Part 1: 36 g (36 capsules) per liter
  • Part 2: potassium phosphate salts and glycerol in sterile solution
  • Contents/l: 24 g yeast extract, 12 g tryptone
  • Powder, part 1 (includes Part 2 sterile liquid)

Terrific broth is a rich nutrient media for propagation of E. coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using Terrific Broth in comparison to traditional LB Broth.

Terrific Broth (TB) has been formulated for optimum growth of E. coli. It is shipped as two separate components, part one contains 24 g Yeast Extract, 12 g Tryptone and part 2 contains Potassium phosphate salts and glycerol in sterile solution.

Capsules: Add 36 capsules to 980 ml of deionized water. Autoclave at 121 °C for 15 minutes. When cool, add 20 ml of sterile Part 2. Powder: Add 36 g to 980 ml of deionized water. Autoclave at 121 °C for 15 minutes. When cool, add 20 ml of sterile Part 2.

Application includes: Bacteria Cell Culture, Gene Expression, Transformation, Microbiological Analysis.

Certifications: Molecular Biology Certified™.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty