Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terrific Broth II (Capsules), MP Biomedicals

Supplier: MP Biomedicals
113046041
76019-964EA 1964.88 USD InStock
76019-964
Terrific Broth II (Capsules), MP Biomedicals
Cell Culture Media
Terrific broth II is a nutritionally rich medium for the growth of E. coli cells containing yeast extract and tryptone in elevated concentrations as well as sodium chloride and vitamins for a luxurious growth.

  • 50 g (50 Capsules) per liter
  • Contents/l: proprietary formulation
  • pH is 6.8 ±0.3 at 25 °C
  • Terrifi c broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components
  • Terrifi c broth II exhibits the same growth characteristics as the original terrifi c broth formula

Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in terrific broth II formulations. Chosen reagents are combined in the accurate proportions according to the formula and thoroughly blended for a uniform distribution. A sample of the blended formulation was used to prepare liquid. The combinations were mixed to dissolve components and autoclaved at 121 °C for 15 minutes.

Terrific broth II is a rich nutrient media for propagation of E. coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using terrific broth in comparison to traditional LB broth.

Terrific broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components. It exhibits the same growth characteristics as the original terrific broth formula.

Add 50 capsules to a liter of purified water and autoclave at 121 °C for 15 minutes.

Application includes: Bacteria Cell Culture, Gene Expression, Transformation, Microbiological Analysis.

Certifications: Molecular Biology Certified™., Gelatin capsules are produced in a facility that meets the FDA’s recent guidance as being derived from BSE-free herds that are born, raised, and slaughtered in the United States.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty