Would you like to log in again?
Pooling Harness Sets, Charter Medical
Supplier: Charter Medical
T8006
MSPP-T8006CS 937.66 USD InStock
MSPP-T8006
Pooling Harness Sets, Charter Medical
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Charter Medical’s disposable multi-lead harness sets are designed for pooling of blood and blood components. Our standard products are suitable for a variety of application like collection, storage, processing, transport, separation, and administration of blood and blood components.
  • Unique shaped spike design
  • Use of pooling harness sets may decrease pool cross contamination
  • Tubing is compatible with sterile connection technology
  • Increase employee safety-spikes not removed from bags
  • Unique spike port provides tamper evident access
  • Decreased product preparation time due to reduced repetitive motions
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty