Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Laurinlactam ≥98%

Supplier: Pfaltz & Bauer
Warning
Synonyms: Azacyclotridecan-2-oneCyclododecanone Isooxime

L01110-100G
101188-312EA 376 USD InStock
101188-312
ω-Laurinlactam ≥98%
omega-Laurinlactam


Formula: C₁₂H₂₃NO
MW: 197.32 g/mol
Boiling Pt: 315 °C
Melting Pt: 149…153 °C
Density: 0,973 g/cm³
Flash Pt: 195 °C
MDL Number: MFCD00005104
CAS Number: 947-04-6
EINECS: 213-424-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty