Would you like to log in again?
MPure™ HPV DNA Extraction Kit for Swab Sample, MP Biomedicals
Supplier: MP Biomedicals
MPure™
117022700
76202-424EA 584.66 USD InStock
76202-424
MPure™ HPV DNA Extraction Kit for Swab Sample, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Nucleic acids extracted from MPure HPV DNA extraction kit for swab samples can be used in a number of downstream applications including: PCR, qPCR, sequencing (NGS), microarray, RFLP and southern blot analysis.
  • Automated nucleic acid purificaton
MPure HPV DNA extraction kit for swab samples is used with the MPure-12 nucleic acid purification system for DNA extraction and purification of the Human Papillomavirus (HPV) from cervical cell samples collected by cervical brush or genital swab in liquid-based medium* (e.g. Hologic Thinprep PreservCyt®, BD Surepath™ and more) or other STM (sample transport media) preservation solutions (e.g. QIAGEN DNA PAP Cervical sampler, Roche Cobas® PCR cell collection media, Hybribio cell preservation solution, and more).
*The liquid-base medium are formulated for cellular preservation and used in liquid-based cytological systems (LBC) for cytological and molecular diagnosis.
Application includes: Nucleic Acid Purification - HPV DNA from swab sample.
Certifications: CE Certified.
Ordering information: Materials required additionally are pipettes, pipette tips, test tubes, gloves.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty