Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl-β-D-xylopyranoside ≥98% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS
Synonyms: 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate

21691-1G 21691-5G
102609-074EA 528.26 USD InStock
102609-074 102598-258
4-Methylumbelliferyl-β-D-xylopyranoside ≥98% (by HPLC)
4-Methylumbelliferyl-beta-D-xylopyranoside


Formula: C₁₅H₁₆O₇
MW: 308.286 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037607
CAS Number: 6734-33-4
Order Now
Specification Test Results
Heavy Metals ≤20 ppm
Water Content (Karl Fisher) ≤1%
Infrared Spectrum Conforms to Structure
Analysis by Mass Spectra Conforms to Standard
4-Methylumbelliferone ≤0.1%
Any Other Impurity ≤0.5%
Total Impurity ≤2%
Residue on Ignition ≤0.5%
¹H NMR Conforms to Structure
Purity by HPLC ≥98%
Purity by TLC ≥98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty