Would you like to log in again?
Shoe Grounder Assembly, SCS, Desco Industries
Supplier: Desco Industries
2051
76238-052EA 46.2 USD InStock
76238-052
Shoe Grounder Assembly, SCS, Desco Industries
Grounders Heel and Toe Grounders
The Shoe Grounder Assembly is designed to effectively ground mobile personnel when they are in contact with conductive and dissipative flooring.
  • Cup style heel grounder with two soles
  • Tab to cup resistance: 1×106 to <1×108 ohms
  • One megohm current limiting resistor
  • Made in the United States of America
The conductive, no-slip outer sole provides a reliable static ground. The function of the shoe grounding assembly depends upon contact with the skin through perspiration in the shoe to sustain an electrical contact between the conductive ribbon and the body. The Shoe Grounder Assembly comes with a 1 Megohm Resistor. A 24" conductive 8-strand nylon polyester grounding tab provides additional conductive material for reliable connection with the operator’s sweat-layer.
Certifications: Meets category ESD SP9.2 Type A & ESD TR53.
Caution: The ESD Series is for electrostatic control. It will not reduce or increase your risk of receiving electric shock when using or working on electrical equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty