Would you like to log in again?
AseptiQuik® S Connectors, Colder Products Company
Supplier: CPC (Colder Products Company)
CPC® AseptiQuik® S
AQS17004HT AQS17002HT
76245-196PK 493.44 USD InStock
76245-196 76245-136
AseptiQuik® S Connectors, Colder Products Company
Fittings and Connectors
Connectors provide quick and easy sterile connections for small-flow applications, even in non-sterile environments.
  • Intuitive three step connection process reduces risk of operator error
  • Repeatable and reliable performance
  • Audible confirmation of assembly
  • Applicable for gamma radiation and autoclave sterilization
  • Temperature range: 4 to 40 °C
The “FLIP-CLICK-PULL” design of AseptiQuik S enables users to quickly transfer small volumes of media easily with less risk of operator error. The connector's genderless and robust design provides reliable performance without the need for clamps, fixtures or tube welders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty