Would you like to log in again?
POLYFLOW II Filter Demi Cartridges, Parker Hannifan
Supplier: Parker
22-MDBMM-100-0
76240-792PK 168.25 USD InStock
76240-792
POLYFLOW II Filter Demi Cartridges, Parker Hannifan
Filters Filter Cartridges
Polyflow II cartridges particularly suitable for the filtration of aggressive and viscous chemicals and solvents.
  • Broad chemical compatibility allows use in most applications
  • Absolute rated beta 10000
High retention efficiency provides excellent protection for downstream filters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty