Would you like to log in again?
Wide Opening Head Vise, PanaVise Products
Supplier: PANAVISE PRODUCTS INC. TE
396
76254-256EA 194.77 USD InStock
76254-256
Wide Opening Head Vise, PanaVise Products
Vises
Ribbed, neoprene jaw pads are 1.875" high, 1.5" wide and provide a sure grip to hold and insulate PC boards and other larger objects; reverse the pads and the wide "V" holds delicate cylindrical objects.
  • Wide opening jaws open to 6" and close to 0"
  • Base accepts all 300 series Heads and any 5/8" diameter shaft
Single knob controls head movement through 3-planes: 90° tilt, 360° turn and 360° rotation.
Base features 4.3125" bolt circle countersunk for three 1/4" flathead screws (not included). Wide opening head vise works perfectly in automotive repair projects and for holding larger objects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty