Would you like to log in again?
Mycophil Agar (LMA), General Laboratory Products
Supplier: General Laboratory Products
250EZ-2100-250
101205-604CS 136.58 USD InStock
101205-604
Mycophil Agar (LMA), General Laboratory Products
Media for Microbiology Prepared Media for Microbiology
LMA (Low pH Mycophil Agar) is used for the enumeration of fungi in sugars, beverages, and other products.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty