Would you like to log in again?
Wisconsin Plankton Sampler
Supplier: SCIENCE INTERACTIVE GROUP
3-40-A50
470308-890EA 402.79 USD InStock
470308-890
Wisconsin Plankton Sampler
Samplers Water Samplers
Sampler enables vertical tows for plankton analysis.
  • Bridle and bucket are stainless steel
A popular and versatile net. The anterior reducing cone makes filtration more efficient than a standard conical net. Sampler consists of an anterior reducing cone; a posterior conical filtering net; and a Dolphin™ adapter with Wisconsin bucket. A stainless steel net ring holds it open where the reducing cone and filter net meet.
Delivery information: Includes matched Wisconsin bucket and adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty