Would you like to log in again?
Luxor Plastic Utility Cart with Tub and Shelves, Market Lab
Supplier: MARKET LAB INC MS
ML14456-BL
MSPP-ML14456BLEA 286.23 USD InStock
MSPP-ML14456BL
Luxor Plastic Utility Cart with Tub and Shelves, Market Lab
Carts
Utility cart with one tub and two shelves.
  • Easily cleanable with standard disinfectants
  • Quiet, easy-rolling casters (locking brakes on 2)
  • Built to handle up to 300 lbs.
Features two flat shelves with a .25" retaining lip and one 2.75" deep tub to prevent items from sliding off.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty