Would you like to log in again?
Plasma/Fluid Transfer Sets, Charter Medical
Supplier: Charter Medical
03-220-90
MSPP-0322090CS 852.18 USD InStock
MSPP-0322090
Plasma/Fluid Transfer Sets, Charter Medical
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
This used for safe and efficient transfer of blood or blood components.
  • Needleless design prevents accidental needle sticks
  • Compatible with sterile connection devices
  • Used for quality control sampling
Attaches to a flexible, non-vented container to prepare, sample, or process blood components.
Aids in compliance with national needle stick safety laws.
Alternative to existing sampling site couplers and potentially hazardous needle/syringe systems.
Packaging: 96 units per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty