Would you like to log in again?
Fluval® Biological Enhancer
Supplier: ROLF C. HAGEN (USA) CORP.
A8346
470308-828EA 2.65 USD InStock
470308-828
Fluval® Biological Enhancer
Animal Supplies Animal Environmental Enrichment
Fluval Biological Enhancer is a responsive biological aquarium supplement that immediately inoculates aquarium water with a powerful team of beneficial bacteria, providing a safe biological habitat for your fish.
  • For fresh and salt water aquariums
  • Powerful team of beneficial bacteria immediately inoculates aquarium water
  • All-natural product
  • Every container contains pure, safe bacterial colonies, free of undesirable contaminants
  • Does not require refrigeration
It goes to work fast, releasing massive amounts of beneficial bacteria that eliminate toxic ammonia and nitrites while creating a biologically well-balanced aquarium for healthy fish to thrive.
Fluval Biological Enhancer is strongly recommended for new aquariums, as they do not contain the necessary concentrations of beneficial bacteria required to metabolize the waste that fish produce. It quickly establishes safe and essential conditions in new aquarium set-ups, so that you can introduce fish to new aquariums immediately.
Regular application helps to competitively exclude the establishment of undesirable bacteria in aquariums. Fluval Biological Enhancer provides consistent results thanks to our unique and proprietary bio-floc technology that ensures product stability, efficacy and shelf life. High-quality production processes ensure that every container contains pure, safe bacterial colonies, free of undesirable contaminants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty