Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Elite® Sponge Filters
Supplier: ROLF C. HAGEN (USA) CORP.
A902 A-900 A905
470308-834EA 15.5 USD InStock
470308-834 470308-836 470014-996
Elite® Sponge Filters
Animal Supplies Animal Environmental Enrichment
A simple and effective filter.
  • Easy operation and maintenance
  • Single airline connection
  • Quiet and safe
The Elite Double BioFoam Sponge Filter is an air-driven filter that efficiently oxygenates and filters aquarium water. The foam filter performs biological and mechanical filtration by trapping floating debris and providing ideal conditions for beneficial bacteria to grow.
To maintain effective biological filtration always rinse the foam filter in water taken from the aquarium. Oxygenation of aquarium water is best achieved by positioning the vertically adjustable outlet at the water surface. This compact internal filter system can conveniently be installed in a variety of locations inside the aquarium and is completely safe for even the smallest baby fish (fry).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty