Would you like to log in again?
Provue Solas XL58 Magnifying Lamp, Aven Tools
Supplier: Aven
ProVue Solas
26501-XL58
76289-848EA 290.44 USD InStock
76289-848
Provue Solas XL58 Magnifying Lamp, Aven Tools
Magnifiers
Equipped with an interchangeable 8-diopter lens.
  • Offers the flexibility of using multiple lenses
  • Fully enclosed spring-balanced arm
  • 60 ultra-bright SMD LEDs with brightness intensity buttons
The Provue Solas XL58 model offers the flexibility of using the included 8-Diopter lens 3× magnification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty