Would you like to log in again?
Marina® 360 Degree Aquarium Kits
Supplier: ROLF C. HAGEN (USA) CORP.
12854
470308-596PK 4.75 USD InStock
470308-596
Marina® 360 Degree Aquarium Kits
Animal Supplies Animal Environmental Enrichment
A full view of your tank with no corners to interfere.
  • Removable cover
  • Custom led lighting
  • Two filter cartridges
  • Three water setup options
"Marina 360 is an innovative desktop aquarium with many functional features designed to make setup and maintenance as easy as possible. The aquarium is uniquely crafted with a sleek housing unit placed discreetly at the back, allowing virtually unobstructed viewing from almost all angles. Small in stature, this 10-litre aquarium occupies minimal space, making it an ideal aquatic showpiece on countertops, desktops and other small spaces, in homes or offices.
Reflecting attention to detail, the plastic edges are light-green tinted to simulate real glass. The cover can be removed completely to provide unobstructed access inside the tank for maintenance. The housing unit-containing a pump, filter and LED lighting-can easily be removed from the tank for maintenance. The pump draws water from the bottom and cycles it back at the top end in a nice gentle flow, providing water circulation at a constant rate of 150 liters per hour.
Two filter cartridges collect organic waste and provide biological filtration to keep the water healthy for fish and plants. They can be pulled out to rinse or replace. The aquarium includes advanced LED lighting that allows you to create a bright daylight setting or blue-tinted nighttime lighting. The aquarium can be set up as a cold, temperate or tropical environment. A heater (sold separately) can be installed in the housing unit.
Ordering information: Gravel and ornaments sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty