Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water nuclease-free

Supplier: New England Biolabs (NEB)
Synonyms: Dihydrogen oxide

B1500L B1500L
103307-278EA 71 USD InStock
CA103307-278 103307-278
Water nuclease-free
Water
Nuclease-free Water is ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in the production of this water, so as to avoid inhibition in enzymatic reactions.

Application: Cellular Analysis, Cloning & Synthetic Biology, DNA Modification
Storage: -20 °C


Formula: H₂O
MW: 18.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty