Would you like to log in again?
Alpha Galactosidase Activity Assay Kit, BioVision
Supplier: BioVision
AB239716-100TEST
76289-474EA 997.29 USD InStock
76289-474
Alpha Galactosidase Activity Assay Kit, BioVision
Assays Enzyme Assays
Alpha galactosidase activity assay kit provides a simple, rapid way to monitor total α-Gal activity in wide variety of biological samples.
  • Quick and easy method
  • Detection limit: <0.1 µU
  • Does not detect beta galactosidase activity
Alpha-Galactosidase (α-Gal; EC 3.2.1.22) hydrolyzes alpha-galactosyl moieties found in glycolipids and glycoproteins. In mammals, α-Gal hydrolyzes poly- and oligosaccharides commonly found in dietary sources that are difficult to digest. Therefore, α-Gal is used in dietary supplements that help to reduce the production of intestinal gases due to consumption of certain foods. It is known total α-Gal activity is due to two major isozymes with unique, yet different thermostability profiles.
Alpha-Galactosidase A, is thermolabile and represents approximately 90% of total α-Gal activity. Fabry Disease, a lysosomal disease disorder, is characterized by mutations in alpha-Galactosidase A. These mutations cause abnormal accumulation of glycosphingolipids in lysosomes.
In this kit, α-Gal cleaves a synthetic specific substrate releasing a fluorophore, which can be easily quantified (Ex/Em= 360/445 nm).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty