Would you like to log in again?
Choline Kinase Activity Assay Kit, BioVision, Inc.
Supplier: BioVision
AB239729-100TEST
76289-478EA 964.45 USD InStock
76289-478
Choline Kinase Activity Assay Kit, BioVision, Inc.
Assays Enzyme Assays
Provides a simple, direct and automation-ready procedure for measuring pyruvate kinase activity in various biological samples such as blood, tissues, and culture cells.
  • Simple procedure, takes ~ less than 40 minutes
  • Fast and convenient
Pyruvate kinase (PK, EC 2.7.1.40) is an enzyme involved in glycolysis. It catalyzes the transfer of a phosphate group from phosphoenolpyruvate (PEP) to ADP, yielding one molecule of pyruvate and one molecule of ATP. Lack of pyruvate kinase will slow down the process of glycolysis which causes the disease known as pyruvate kinase deficiency.
In the assay, PEP and ADP were catalyzed by PK to generate pyruvate and ATP. The generated pyruvate is oxidized by pyruvate oxidase to produce color (at λ = 570 nm) and fluorescence (at Ex/Em = 535/587 nm). Since the increase in color or fluorescence intensity is proportional to the increase in pyruvate amount, the PK activity can be accurately measured. The kit detects 0.1 mU/ml pyruvate kinase.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty