Would you like to log in again?
PicoProbe™ Hypoxanthine Phosphoribosyl Transferase Activity Assay Kit, BioVision, Inc.
Supplier: BioVision
AB284551-100TEST
76289-480EA 895.44 USD InStock
76289-480
PicoProbe™ Hypoxanthine Phosphoribosyl Transferase Activity Assay Kit, BioVision, Inc.
Assays Enzyme Assays
HPRT activity assay kit is a simple one-step plate based assay kit for the measurement of HPRT activity in biological samples.
  • Simple, rapid and convenient assay to measure HPRT activity in purified enzyme
  • Includes positive control
  • Detection limit: 2 mU
Hypoxanthine Phosphoribosyl Transferase or HPRT (EC 2.4.2.8) plays an important role in the generation of purine nucleotides through the purine salvage pathway and is present in prokaryotes and eukaryotes. The enzyme converts guanine to guanosine monophosphate and hypoxanthine to inosine monophosphate by transferring a phosphoribosyl group from phosphoribosyl pyrophosphate (PRPP) to the purine molecule, and releases an inorganic pyrophosphate molecule in the process.
Mutations in HPRT lead to developmental disorders such as Lesch Nyhan Disease (LND), characterized by neurological and behavioral abnormalities. Its deficiency also leads to defective basal ganglia expression of the neurotransmitter dopamine and aberrant neuronal function. This causes dysregulation of multiple dopamine- related developmental functions and cellular signaling defects.
HPRT catalyzes the conversion of hypoxanthine to inosine monophosphate (IMP). IMP then undergoes a series of enzymatic reactions to convert a non-fluorescent probe to a fluorescent product. The signal, which is proportional to the generated product can be read in kinetic mode at Ex/Em 535/587 nm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty