Would you like to log in again?
Phosphoglycerate Dehydrogenase Activity Assay Kit, BioVision, Inc.
Supplier: BioVision
AB273328-100TEST
76289-466EA 1055.59 USD InStock
76289-466
Phosphoglycerate Dehydrogenase Activity Assay Kit, BioVision, Inc.
Assays Enzyme Assays
Phosphoglycerate Dehydrogenase assay kit provides a quick and easy method for monitoring PHGDH activity in a variety of samples.
  • Simple, rapid and convenient assay to measure PHGDH activity in biological samples
  • High–throughput adaptable
  • The kit can detect PHGDH activity less than 0.1 mU per sample
Phosphoglycerate Dehydrogenase (PHGDH: EC 1.1.1.95) is the first enzyme that participates in the L-serine synthesis pathway. PHGDH catalyzes the reversible reaction that converts 3-phosphoglycerate into 3-phosphohydroxyglycerate and the NADH-dependent reduction of α-ketoglutarate into D-2-hydroxyglutarate (D-2HG).
In humans, PHGDH deficiency causes severe health problems including congenital microcephaly, psychomotor retardation, and intractable seizure. Recent studies have shown increased PHGDH activity in a wide variety of cancers, particularly in breast and melanoma tumors. Therefore, the measurement of PHGDH activity has become of great importance in searching potential therapeutic agents against cancer.
In this kit, Phosphoglycerate Dehydrogenase converts 3-phosphoglycerate and NAD into 3-phosphohydroxyglerate and NADH respectively. The oxidation of NADH reduces a probe generating a strong, stable absorbance signal (OD: 450 nm).
Delivery information: Includes positive control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty