Would you like to log in again?
EZScreen™ Alanine Aminotransferase Activity Assay Kit, BioVision, Inc.
Supplier: BioVision
AB241035-400TEST
76289-484EA 865.97 USD InStock
76289-484
EZScreen™ Alanine Aminotransferase Activity Assay Kit, BioVision, Inc.
Assays Enzyme Assays
The assay is suitable for high throughput activity of ALT with a detection limit of 10 mU per well.
  • Simple, fast and convenient procedure
  • Sensitive assays for measuring ALT activity in various biological samples and the kit is also suitable for high throughput analyses
  • Detection limit: 10 mU per well
Alanine transaminase (ALT) is a transaminase enzyme (EC 2.6.1.2). It is also called serum glutamic pyruvic transaminase (SGPT) or alanine aminotransferase (ALAT). ALT is found in serum and in various bodily tissues, but is usually associated with the liver. It is commonly measured clinically as a part of a diagnostic liver function test to determine liver health. Diagnostically, it is almost always measured in units/liter (U/L).
It catalyzes the reaction: α-ketoglutarate + alanine ⇌ glutamate + pyruvate.
ALT assay Kit, ALT catalyzes the transfer of an amino group from alanine to ⍺-ketoglutarate, the products of this reversible transamination reaction being pyruvate and glutamate. The pyruvate is detected in a reaction that concomitantly converts a nearly colorless probe to both color (λmax = 570 nm) and fluorescence (Ex/Em = 535/587 nm).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty