Would you like to log in again?
PicoProbe™ Glucokinase Activity Assay Kit, BioVision, Inc.
Supplier: BioVision
AB273303-100TEST
76289-486EA 997.97 USD InStock
76289-486
PicoProbe™ Glucokinase Activity Assay Kit, BioVision, Inc.
Assays Enzyme Assays
PicoProbe™ glucokinase activity assay kit provides a quick and easy method for monitoring GCK activity in wide variety of samples.
  • Simple and rapid procedure
  • Highly sensitive
  • The assay is sensitive, specific, and high-throughput adaptable
  • It can detect as low as 2 µU of GCK activity
Glucokinase (also called GCK, hexokinase type IV or D and ATP: D-hexose 6-phosphotransferase; EC 2.7.1.1) is expressed in specific types of tissues: liver, pancreas, small intestine and brain. Glucokinase functions as a glucose sensor, triggering shifts in carbohydrate metabolism or cell function in response to the levels of glucose in blood, such as nutritional and hormonal molecular pathways. Unlike other Hexokinases, Glucokinase has a relatively low affinity for glucose and it is not inhibited by physiological concentrations of glucose 6-phosphate.
Mutations in the gene encoding GCK can cause both hyperglycemia and hypoglycemia. Due to the major role of Glucokinase in controlling blood glucose homeostasis, Glucokinase is currently considered as a strong candidate target for the treatment of Hyperglycemia, a condition encountered in Type 2 Diabetic patients.
In this assay, GCK converts glucose into glucose-6-phosphate, which in turn is converted into a series of intermediates that reduce PicoProbeTM generating an intense fluorescence product (Ex/Em=535/587 nm).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty