Would you like to log in again?
Total Iron-Binding Capacity and Serum Iron Assay Kit, BioVision
Supplier: BioVision
AB239715-100TEST
76289-472EA 948.88 USD InStock
76289-472
Total Iron-Binding Capacity and Serum Iron Assay Kit, BioVision
Assays Cellular Assays
TIBC and serum iron assay kit measures both total iron-binding capacity and serum iron.
  • Simple procedure, takes ~ 1 hour
  • Fast and convenient
  • The assay is sensitive, stable and high-throughput adaptable
  • Useful for the estimation of TIBC, unbound iron, transferrin saturation, serum iron
Those values indicate the requisite iron for transferrin saturation and serum iron respectively. In humans, transferrin is a blood protein that binds and transports iron throughout the body.
Measurements can be used for to detect and monitor transferrin saturation and also iron-deficiency anemia and chronic inflammatory diseases.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty