Would you like to log in again?
PicoProbe™ Methylglyoxal Assay Kit, BioVision
Supplier: BioVision
AB273284-100TEST
76289-476EA 965.6 USD InStock
76289-476
PicoProbe™ Methylglyoxal Assay Kit, BioVision
Assays Cellular Assays
The fluorometric assay kit that can detect methylglyoxal in samples.
  • This kit provides a simple, reproducible and highly sensitive in vitro assay for detection
  • Detection limit: 6 pmol of MG in a 100 µl reaction
Methylglyoxal (MG, 2-oxopropanal, pyruvic aldehyde), a by-product of glycolysis, is a highly toxic, reactive dicarbonyl compound. It causes non-enzymatic glycation of proteins which yields irreversible advanced glycation end products (AGEs), leading to cross-linking or degradation of proteins. MG is associated with several pathologies including diabetes, aging and neurodegenerative diseases. Furthermore, recent studies have suggested that MG may induce carbonyl stress-type schizophrenia.
PicoProbe™ methylglyoxal assay kit (Fluorometric) can detect MG using an enzyme-coupled reaction which reduces the fluorogenic probe. The reduced fluorophore produces a stable signal (Ex/Em= 535/587nm), which is directly proportional to the amount of MG in samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty