Would you like to log in again?
Methylglyoxal Assay Kit, BioVision
Supplier: BioVision
AB241006-100TEST
76289-464EA 909.52 USD InStock
76289-464
Methylglyoxal Assay Kit, BioVision
Assays Cellular Assays
The assay kit used to detect Methylglyoxal in samples.
  • Simple procedure, takes ~ 2 hours
  • Fast and convenient
  • The assay is sensitive, stable and high-throughput adaptable
  • Useful for the estimation of methylglyoxal in biological samples
Methylglyoxal (MG, 2-oxopropanal, pyruvic aldehyde) is a highly reactive dicarbonyl compound formed during glycolysis and sugar fermentation. It is an important precursor of advanced glycation end products (AGEs). Endogenous MG is present in many food items and beverages. For example, high levels of MG have been reported in Manuka Honey and some soft drinks.
Additionally, cooking and some storage techniques can induce the synthesis of MG, such as coffee brewing, and bread toasting. It has been shown that MG has antimicrobial activity against some bacterial strains, including Streptococcus, H. pylori, E. coli and other bacteria resistant to standard antibiotic treatments.
Methylglyoxal Assay Kit (Colorimetric) enables the detection of MG using a set of engineered enzymes and a chromophore. The reduced chromophore, final product of the assay, produces a stable signal, which can be easily quantified at 450 nm using a microplate reader and its signal is directly proportional to the amount of MG in samples. The assay is simple, specific, reproducible, and can detect as low as 0.5 nmol/well of MG in a 100 µl reaction.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty