Would you like to log in again?
Pepsin/Pepsinogen Activity Assay Kit, BioVision
Supplier: BioVision
AB239722-100TEST
76289-460EA 712.71 USD InStock
76289-460
Pepsin/Pepsinogen Activity Assay Kit, BioVision
Assays Enzyme Assays
Pepsin assay kit is a homogenous assay that allows for quantification of pepsin activity in gastric tissue and various biological fluids.
  • Simple and highly sensitive procedure, takes ~1 to 2 hours based on pepsin levels
  • Fast and convenient
  • Detection limit: 550 μU
Pepsin (EC 3.4.23.1) is a peptidase A1 family aspartic protease that serves as a primary digestive enzyme in the mammalian stomach. Pepsin is synthesized in the chief cells of the gastric mucosa as the inactive pro-enzyme pepsinogen, which contains a 44 amino acid autoinhibitory domain. The proenzyme is packaged into granules and stored inside the gastric mucosal cells. When food is consumed, neuronal signals traveling along the vagus nerve trigger secretion of HCl and pepsinogen into the stomach. In the highly acidic environment of the stomach, pepsinogen unfolds and undergoes autocatalytic cleavage, forming the active protease pepsin. Infection by the bacterium H. pylori induces hypersecretion of both stomach acid and pepsin, contributing to the formation of gastric and duodenal ulcers.
In addition, excessive amounts of pepsin can play a role in the degradation of the esophageal and laryngeal tissue observed in chronic gastroesophageal reflux disease. While the majority of pepsinogen is expressed in the gastric mucosa, a small amount of the inactive enzyme is also released into the blood. Serum pepsinogen levels correlate with functional changes to gastric mucosa and are often measured for the screening of gastric cancer and chronic H. pylori induced gastritis.
The assay utilizes a synthetic peptide substrate bearing both a fluorophore and a fluorescence quencher. Upon cleavage by pepsin, the fluorophore-bearing peptide fragment is unquenched to produce a bright fluorescent signal (Ex/Em = 328/418 nm). Lysosomal aspartic proteases in the peptidase A1 family (Cathepsin D and E) do not interfere with the assay.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty