Would you like to log in again?
Distillation Adapter, with Two [ST] Joints, Drip Tip, and Hose Connection, Chemglass
Supplier: Chemglass
CG-1072-B-14
80066-172EA 220.12 USD InStock
80066-172
Distillation Adapter, with Two [ST] Joints, Drip Tip, and Hose Connection, Chemglass
Distillation Components Distillation Adapters
With two standard taper inner joints, parallel, at a 75° angle to the connecting tube.
  • 14/20 Joint
  • 100 mm center to center
  • 75° angle to the connecting tube
Adapter has a sealed-in drip tip and serrated hose connection for vacuum source.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty