Would you like to log in again?
EZBlock™ Protease Inhibitor Cocktail XVI, Biovision
Supplier: BioVision
AB289878-1VIAL
76290-152EA 152.98 USD InStock
76290-152
EZBlock™ Protease Inhibitor Cocktail XVI, Biovision
Cell Culture Supplements and Additives
A Protease inhibitor cocktail
A cocktail consisting of five protease inhibitors useful for the inhibition of aspartic, cysteine, serine, metalloproteases and aminopeptidases. Each vial should be reconstituted with 1 ml DMSO and 4 ml H₂O to obtain a 5 ml stock solution. This protease inhibitor cocktail is recommended for use with bacterial cell extracts (except in case of use in metal ion chromatography). One set is sufficient for about 20 grams of cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty