Would you like to log in again?
Sampling Adapters, 24/40, Chemglass
Supplier: Chemglass
CG-1037-20 CG-1037-10 CG-1037-16 CG-1037-11 CG-1037-12 CG-1037-15
16002-612EA 259.43 USD InStock
16002-612 16002-610 16002-616 16002-614 16002-608 16002-618
Sampling Adapters, 24/40, Chemglass
Adapters Adapters, Joints with Tube Connectors
Sampling adapter for use with a 24/40 outer joint on the reaction kettle lid.
  • Complete adapter supplied with 3-port 24/40 PTFE adapter, and 10' of 1/8" O.D. PTFE tubing
  • Adapter and valve have 1/4" – 28 female threads
  • Adapter has three inlets for use with 1/8" O.D. PTFE tubing or optional 5 mm O.D. silicone septa
Adapter is supplied complete with a 90° bore valve enabling connection to a luer-type syringe for product sampling.
Delivery information: Adapter is supplied complete with (3) nuts, (3) ferrules, and (2) solid plugs. Sampling valve is also supplied complete with nuts and ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty