Would you like to log in again?
Airfree® Schlenk Solvent Transfer Manifolds, Chemglass
Supplier: Chemglass
Airfree®
AF-0558-01
80064-968EA 436.08 USD InStock
80064-968
Airfree® Schlenk Solvent Transfer Manifolds, Chemglass
Manifolds
Designed to purify and transfer solvents under inert conditions.
  • Equipped with a Chem-Vac™ Chem-Cap®
  • With 24/40 joint
Manifold permits transfer of solvents directly into an evacuated storage vessel with 24/40 standard taper outer joints. A 0 to 4 mm Chem-Vac™ Chem-Cap® valve is used for vacuum control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty