Would you like to log in again?
Proto® Pin Punch Sets, ORS Nasco
Supplier: ORS Nasco
577-47A-TT
76190-706EA 411.91 USD InStock
76190-706
Proto® Pin Punch Sets, ORS Nasco
Tool Sets
A polished identification panel on one of the flats is marked with the tool number and size in a black inlay.
  • Made from S2 steel and specially heat-treated for greater strength and durability
  • Two-piece design allows the tip to be hard for durability while allowing it to be produced straight to help prevent binding
  • Modified parabolic striking end to help reduce mushrooming and chipping
  • Black oxide finish to help prevent rust corrosion, extending tool life and performance
Delivery information: Includes a pouch for convenient storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty