Would you like to log in again?
Proto® Crowfoot Wrenches, Essendant
Supplier: Stanley
577-5320MCF
76189-468EA 73.69 USD InStock
76189-468
Proto® Crowfoot Wrenches, Essendant
Wrenches
Anti-Slip Design open end (ASD) has locking grooves that help prevent the wrench from slipping off the fastener when turning.
  • Crowfoot wrenches are used in hard-to-reach, restricted access situations where a standard wrench or ratchet cannot reach
  • Full polish finish provides corrosion resistance and easy-to-clean surfaces
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty