Would you like to log in again?
Proto® Torque Multipliers, Essendant
Supplier: Stanley
577-6212CERT
76189-732EA 2795.29 USD InStock
76189-732
Proto® Torque Multipliers, Essendant
Wrenches
  • Use with torque wrenches and heavy-duty power or impact sockets
  • Tubular reaction bar
  • Single stage planetary gear system
  • Standard torque accuracy is ±5% of reading
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty