Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Drive Universal Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-77274P
76189-832EA 53.27 USD InStock
76189-832
Proto® Drive Universal Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
  • Ball and spring design allows for a 30° working angle
  • Smooth 360° rotation
  • Ideal for use on stubborn or damaged fasteners, or fasteners made from soft materials
The TorquePlus™ configuration employs a radius-corner design, shifting contact area and stress away from the fastener corners and distributing contact stresses over a large area of the socket, minimizing slippage and fastener rounding, helping to extend fastener life, and allowing the user to more easily turn worn, rounded, or rusted fasteners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty