Would you like to log in again?
Proto® DuraTek™ Slotted Square Bar Screwdrivers, ORS Nasco
Supplier: ORS Nasco
577-K51606S
76191-056EA 21.21 USD InStock
76191-056
Proto® DuraTek™ Slotted Square Bar Screwdrivers, ORS Nasco
Screwdrivers
Designed to meet demanding heavy-duty needs of industrial maintenance and assembly applications.
  • Ergonomic quad-lobe handle design with 360° texture for additional grip
  • Impact and chemical resistant handles are four-sided with broad but shallow flutes
  • Resistant to most harsh chemicals
  • Fully grounded and chemically polished, then chrome plated for corrosion resistance
  • Tip is made of high-alloy steel
Resistant to most harsh chemicals (including hydraulic fluids used in the aerospace industry). Four-sided handle design helps prevent "roll away" from the work surface. High-alloy steel shanks are heat-treated their entire length to help provide strength against twisting action and to help prevent bending and twisting when placed under torque. Precision machined for precise fastener fit. Finished in black oxide to help prevent cam out and slipping. Made in USA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty