Would you like to log in again?
Proto® Leverage Wrenches, Essendant
Supplier: Stanley
577-2650PW 577-2617PW 577-2632PW
76188-712EA 410.12 USD InStock
76188-712 76188-668 76190-636
Proto® Leverage Wrenches, Essendant
Wrenches
Leverage wrenches provide a safe alternative to striking wrenches in confined spaces and on applications that prohibit sparking.
  • Offset box wrench head offers access to recessed fasteners and keeps hands clear of obstructions
  • Ergonomic design allows wrenches to be used conventionally without the handle
  • The handle can be attached to provide increased leverage for final tightening
  • Bar stop on the wrench shaft helps to align locking button with handle hole
  • Ideal for oil and gas industries
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty