Would you like to log in again?
Proto® Split Rings, ORS Nasco
Supplier: ORS Nasco
577-SR15 577-SR2 577-SR1 577-SR04
76190-356PK 65.54 USD InStock
76190-356 76190-346 76190-350 76190-354
Proto® Split Rings, ORS Nasco
Tool Sets
Installs easily to manufacturers pre-drilled tether holes.
  • Fits tools of significant geometry
  • Create a tether ring on the fly
  • Weight rated for a max tool weight of 2 lbs.
  • Semi-permanent attachment of tools to lanyards
  • For use with tethers 12" or less
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty