Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® C-Clamp Forcing Screws, Stanley® Products
Supplier: Stanley
577-1P1-HD 577-3P2-HD 577-10P2-HD 577-11/2P1-HD
76188-578EA 29.43 USD InStock
76188-578 76190-670 76188-574 76190-542
Proto® C-Clamp Forcing Screws, Stanley® Products
Clamps General Purpose Clamps
Full-length forcing screws provide greater versatility with the minimum capacity always being "zero".
  • Strongest C-clamp design
  • Heavy-duty threaded hub provides maximum screw support
  • Square head
  • Hardened tip
Forcing screws have a square head that is designed to be turned with a wrench and a hardened tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty