Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proto® Tether-Ready Heavy-Duty Pistol Grip Grease Guns, Stanley® Products

Supplier: Stanley
577-FC33HD-TT
76189-994EA 141.77 USD InStock
76189-994
Proto® Tether-Ready Heavy-Duty Pistol Grip Grease Guns, Stanley® Products
Spray Guns
Tether-ready with heat shrink loop attachment with three-way filling: bulk, cartridge, or filler pump.

  • 16 oz. bulk fill, 14 oz. standard cartridge
  • Heavy-duty 1.7 mm thick seamless barrel to avoid dents
  • Aluminum head reduces weight
  • Heavy-duty flexible 18" nylon hose and 6" metal spout for easy use
  • Output pressure: 3600 PSI

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty