Would you like to log in again?
LaMotte® Digital Tapwater Tour
Supplier: LaMotte
5972 R-5972
470308-014EA 86 USD InStock
470308-014 470308-012
LaMotte® Digital Tapwater Tour
Educational Materials Environmental Educational Materials Environmental Learning Activities
What's coming out of your tap?
  • Perfect for grades 4 to 8
  • Aligned to NGSS 5-ESS3-1, MS-PS1-2, MS-ESS3-1, MS-ESS3-4, HS-ESS2-5, HS-ESS3-1
  • Instructor manual with everything you need from background information and setting up the classroom to a shopping list for everyday items
  • Student manual including test procedure, data sheets, and puzzles
  • Encourages critical thinking and active learning
  • Works on computer or tablet
  • Great for inclusion classrooms
The newly updated electronic version of our ever popular Tapwater Tour® is a classroom investigation of how and why the characteristics of tap water can vary and aff ect every day activities. After reviewing background material about pH, chlorine, hardness, copper, and iron, students work independently and in teams to test their own tap water samples with simple TestTabs® reagent tablets. Students use observations and analyze shared data from the hands-on activities to explore water quality. Activities can be modified to fit specific curriculum needs or learning opportunities. Data sheets and puzzles for each unit can be printed or filled electronically by students using a computer or tablet.
Delivery information: Includes instructor manual, student manual with data sheets and puzzles, classroom color chart, and reagents and sample test bags for 50 students.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty