Would you like to log in again?
Adapters, Sanitary to Standard Taper, Glass Filled PTFE, Tri-Clamp, Chemglass
Supplier: Chemglass
CG-1002-B-12 CG-1002-B-10
89492-738EA 243.24 USD InStock
89492-738 89492-740
Adapters, Sanitary to Standard Taper, Glass Filled PTFE, Tri-Clamp, Chemglass
Adapters Adapters, Conical Ground Joints
Provides a convenient way to connect sanitary fittings and adapters to chemglass reaction systems.
  • Overall length is 4" (102 mm)
  • Glass-filled PTFE
Standard taper inner joint to sanitary fitting adapter.
Ordering information: Clamp and seals must be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty