Would you like to log in again?
Heavy-Duty, Non-Magnetic #304 Stainless Steel Spatula, Chemglass
Supplier: Chemglass
CG-1986-20
16003-202EA 71.64 USD InStock
16003-202
Heavy-Duty, Non-Magnetic #304 Stainless Steel Spatula, Chemglass
Spatulas
Heavy-duty, extra-long spatula is made from non-magnetic stainless steel.
  • One end is rounded, the other is flat
  • Non-magnetic
  • Stainless steel construction
Spatula will pass through a 24/40 outer joint. Both ends are ³/₄" (19 mm)×3" (75 mm) (Ø×L)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty