Would you like to log in again?
Tefzel® Flangeless Ferrules, Chemglass
Supplier: Chemglass
CG-1164-F-02
80060-478EA 4.93 USD InStock
80060-478
Tefzel® Flangeless Ferrules, Chemglass
Fittings and Connectors
Ferrules are designed to form a zero dead-volume connection.
  • Use with flangeless nut to ensure tubing is held firmly in place
  • Yellow finish
Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty