Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RN 1 dihydrochloride

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: 1-(4-methyl-1-piperazinyl)-2-[[(1R*,2S*)-2-[4-phenylmethoxy)phenyl]cyclopropyl]amino]ethanone dihydrochloride

AB254240-25MG AB254240-5MG
76289-968EA 1170.31 USD InStock
76289-968 76289-966
RN 1 dihydrochloride
RN 1 dihydrochloride
A cell-permeable, lysine specific demethylase 1 (LSD1) inhibitor

RN 1 dihydrochloride is a potent, cell-permeable, lysine specific demethylase 1 (LSD1) inhibitor (IC₅₀ = 70 nM). Displays selectivity for LSD1 over MAO-A and MAO-B (IC₅₀ values are 0.51 and 2.79 μM for MAO-A and MAO-B, respectively). Impairs long-term, but not short-term memory in mice.


Formula: C₂₃H₂₉N₃O₂.2HCl
MW: 452,42 g/mol
Storage Temperature: Freezer
CAS Number: 1781835-13-9
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty